Tisk…

1'-(5-Bromo-6-chloro-4-(trifluoromethyl)pyridin-2-yl)piperazine hydrobromide

Dodavatel: Apollo Scientific

PC446019-1G PC446019-5G
APOSPC446019-1GEA 6950 CZK
APOSPC446019-1G APOSPC446019-5G
1'-(5-Bromo-6-chloro-4-(trifluoromethyl)pyridin-2-yl)piperazine hydrobromide
1'-(5-Bromo-6-chloro-4-(trifluoromethyl)pyridin-2-yl)piperazine hydrobromide
Vzorec: C₁₀H₁₁Br₂ClF₃N₃
Storage Temperature: Ambient
MDL Number: MFCD11052384
CAS číslo: 1208081-40-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR