Tisk…

N'-[2-(4-Chlorophenyl)-1-(2,2,2-trifluoroethylamino)ethylidene]hydrazinecarboxylic acid-tert-butyl

Dodavatel: Apollo Scientific

PC446012-1G PC446012-500MG
APOSPC446012-1GEA 5200 CZK
APOSPC446012-1G APOSPC446012-500MG
N'-[2-(4-Chlorophenyl)-1-(2,2,2-trifluoroethylamino)ethylidene]hydrazinecarboxylic acid-tert-butyl
N'-[2-(4-Chlorophenyl)-1-(2,2,2-trifluoroethylamino)ethylidene]hydrazinecarboxylic acid-tert-butyl
Vzorec: C₁₅H₁₉ClF₃N₃O₂
MW: 365,79 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568263
CAS číslo: 1053658-11-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR