N'-[2-Phenoxy-1-(2,2,2-trifluoroethylamino)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
Dodavatel: Apollo Scientific
PC446011-1G
PC446011-500MG
APOSPC446011-1GEA
5200
CZK
APOSPC446011-1G
APOSPC446011-500MG
N'-[2-Phenoxy-1-(2,2,2-trifluoroethylamino)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[2-Phenoxy-1-(2,2,2-trifluoroethylamino)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
Vzorec:
C₁₅H₂₀F₃N₃O₃ MW: 347,34 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10568260 CAS číslo: 1053657-91-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...