N'-[1-Amino-1-(3-fluoro-4-methylphenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
Dodavatel: Apollo Scientific
PC446005-1G
PC446005-500MG
APOSPC446005-1GEA
5200
CZK
APOSPC446005-1G
APOSPC446005-500MG
N'-[1-Amino-1-(3-fluoro-4-methylphenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-1-(3-fluoro-4-methylphenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
Vzorec:
C₁₃H₁₈FN₃O₂ MW: 267,31 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10568192 CAS číslo: 1053655-97-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...