N'-[1-Amino-1-(3,5-bis-(trifluoromethyl)phenyl)methylidene]hydrazine carboxylic acid-tert-butyl ester
Dodavatel: Apollo Scientific
PC446004-1G
PC446004-500MG
APOSPC446004-1GEA
5200
CZK
APOSPC446004-1G
APOSPC446004-500MG
N'-[1-Amino-1-(3,5-bis-(trifluoromethyl)phenyl)methylidene]hydrazine carboxylic acid-tert-butyl ester
N'-[1-Amino-1-(3,5-bis-(trifluoromethyl)phenyl)methylidene]hydrazine carboxylic acid-tert-butyl ester
Vzorec:
C₁₄H₁₅F₆N₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD10568191 CAS číslo: 1053655-95-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...