N'-[1-Amino-1-(4-(trifluoromethoxy)phenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
Dodavatel: Apollo Scientific
PC446001-1G
PC446001-500MG
APOSPC446001-1GEA
5200
CZK
APOSPC446001-1G
APOSPC446001-500MG
N'-[1-Amino-1-(4-(trifluoromethoxy)phenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-1-(4-(trifluoromethoxy)phenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
Vzorec:
C₁₃H₁₆F₃N₃O₃ MW: 319,29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10568179 CAS číslo: 1053655-71-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...