N'-[1-Amino-1-(3-(trifluoromethyl)phenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
Dodavatel: Apollo Scientific
PC446000-1G
PC446000-500MG
APOSPC446000-1GEA
5200
CZK
APOSPC446000-1G
APOSPC446000-500MG
N'-[1-Amino-1-(3-(trifluoromethyl)phenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-1-(3-(trifluoromethyl)phenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
Vzorec:
C₁₃H₁₆F₃N₃O₂ MW: 303,29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10568174 CAS číslo: 1053655-65-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...