Tisk…

4,5,5-Trifluoropent-4-enoic acid 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC4456-1G PC4456-5G
APOSPC4456-1GEA 2820 CZK
APOSPC4456-1G APOSPC4456-5G
4,5,5-Trifluoropent-4-enoic acid 97%
4,5,5-Trifluoropent-4-enoic acid
Vzorec: C₅H₅F₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00156072
CAS číslo: 110003-22-0
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR