Tisk…

5,5,5-Trifluoropentanoic acid 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC4455-1G PC4455-250MG PC4455-5G
APOSPC4455-1GEA 1820 CZK
APOSPC4455-1G APOSPC4455-250MG APOSPC4455-5G
5,5,5-Trifluoropentanoic acid 97%
5,5,5-Trifluoropentanoic acid
Vzorec: C₅H₇F₃O₂
Bod varu: 94…95 °C/15mmHg
Hustota: 1,293 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02262192
CAS číslo: 407-62-5
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR