Tisk…

2-Fluoro-4-iodo-6-methylpyridine 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Fluoro-4-iodo-6-picoline

PC445040-1G PC445040-250MG
APOSPC445040-1GEA 13460 CZK
APOSPC445040-1G APOSPC445040-250MG
2-Fluoro-4-iodo-6-methylpyridine 95%
2-Fluoro-4-iodo-6-methylpyridine
Vzorec: C₆H₅FIN
Storage Temperature: Ambient
MDL Number: MFCD03095298
CAS číslo: 884494-45-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR