Tisk…

5-Fluoro-2-nitropyridine

Dodavatel: Apollo Scientific
Varování

PC445034-250MG PC445034-1G PC445034-5G
APOSPC445034-250MGEA 1240 CZK
APOSPC445034-250MG APOSPC445034-1G APOSPC445034-5G
5-Fluoro-2-nitropyridine
5-Fluoro-2-nitropyridine
Vzorec: C5H3FN2O2
MW: 142,0879 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03092917
CAS číslo: 779345-37-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR