Tisk…

Gadolinium trifluoride 99,9%

Dodavatel: Apollo Scientific

Synonyms: Gadolinium (III) fluoride, Gadolinium(III) bromide

PC4386P-100G PC4386P-50G PC4386P-10G
APOSPC4386P-100GEA 9530 CZK
APOSPC4386P-100G APOSPC4386P-50G APOSPC4386P-10G
Gadolinium trifluoride 99,9%
Gadolinium trifluoride
Vzorec: F₃Gd
MW: 214.25
Bod tání: 1231
Storage Temperature: Ambient
MDL Number: MFCD00016113
CAS číslo: 13765-26-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR