5-Fluorouridine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC4380B-1G
PC4380B-5G
APOSPC4380B-1GEA
1780
CZK
APOSPC4380B-1G
APOSPC4380B-5G
5-Fluorouridine 97%
5-Fluorouridine
Vzorec:
C₉H₁₁FN₂O₆ Bod tání: 182…185 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00036832 CAS číslo: 316-46-1 EINECS: 206-260-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...