Tisk…

4-Fluoro-2-(trifluoromethyl)benzyl bromide 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC4376J-10G PC4376J-100G PC4376J-50G
APOSPC4376J-10GEA 622 CZK
APOSPC4376J-10G APOSPC4376J-100G APOSPC4376J-50G
4-Fluoro-2-(trifluoromethyl)benzyl bromide 98%
4-Fluoro-2-(trifluoromethyl)benzyl bromide
Vzorec: C₈H₅BrF₄
Storage Temperature: Ambient
MDL Number: MFCD00061176
CAS číslo: 206860-48-2
UN: 1760
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR