Tisk…

4-Fluoro-3-(trifluoromethyl)benzyl alcohol 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-Fluoro-3-(trifluoromethyl)phenyl)methanol

PC4375K-1G PC4375K-5G PC4375K-25G
APOSPC4375K-1GEA 622 CZK
APOSPC4375K-1G APOSPC4375K-5G APOSPC4375K-25G
4-Fluoro-3-(trifluoromethyl)benzyl alcohol 97%
4-Fluoro-3-(trifluoromethyl)benzyl alcohol
Vzorec: C₈H₆F₄O
MW: 194.13
Bod varu: 80…85 °C/0.1mmHg
Storage Temperature: Ambient
MDL Number: MFCD00061171
CAS číslo: 67515-61-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR