Tisk…

3-Fluoro-5-(trifluoromethyl)benzyl alcohol 97%

Dodavatel: Apollo Scientific
Varování

PC4375I-1G PC4375I-10G PC4375I-5G PC4375I-25G
APOSPC4375I-1GEA 622 CZK
APOSPC4375I-1G APOSPC4375I-10G APOSPC4375I-5G APOSPC4375I-25G
3-Fluoro-5-(trifluoromethyl)benzyl alcohol 97%
3-Fluoro-5-(trifluoromethyl)benzyl alcohol
Vzorec: C₈H₆F₄O
MW: 194,13 g/mol
Bod varu: 80…82 °C/3mmHg
Storage Temperature: Ambient
MDL Number: MFCD00061169
CAS číslo: 184970-29-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR