Tisk…

2-Fluoro-6-(trifluoromethyl)benzyl alcohol 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (2-Fluoro-6-(trifluoromethyl)phenyl)methanol

PC4375H-1G PC4375H-25G PC4375H-5G
APOSPC4375H-1GEA 622 CZK
APOSPC4375H-1G APOSPC4375H-25G APOSPC4375H-5G
2-Fluoro-6-(trifluoromethyl)benzyl alcohol 97%
2-Fluoro-6-(trifluoromethyl)benzyl alcohol
Vzorec: C₈H₆F₄O
MW: 194,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061168
CAS číslo: 152211-15-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR