Tisk…

4-Fluoro-3-(trifluoromethyl)benzylamine 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC4374Z-25G PC4374Z-5G
APOSPC4374Z-25GEA 9660 CZK
APOSPC4374Z-25G APOSPC4374Z-5G
4-Fluoro-3-(trifluoromethyl)benzylamine 97%
4-Fluoro-3-(trifluoromethyl)benzylamine
Vzorec: C₈H₇F₄N
Bod varu: 101 °C/3mmHg
Storage Temperature: Ambient
MDL Number: MFCD00070814
CAS číslo: 67515-74-6
UN: 2735
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR