2-Fluoro-4-(trifluoromethyl)benzylamine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC4374U-500MG
PC4374U-250MG
APOSPC4374U-500MGEA
705
CZK
APOSPC4374U-500MG
APOSPC4374U-250MG
2-Fluoro-4-(trifluoromethyl)benzylamine 97%
2-Fluoro-4-(trifluoromethyl)benzylamine
Vzorec:
C₈H₇F₄N Storage Temperature: Ambient |
MDL Number:
MFCD00070807 CAS číslo: 239087-05-9 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...