4-Fluoro-3-(trifluoromethyl)benzamide 98%
Dodavatel: Apollo Scientific
PC4371X-25G
PC4371X-5G
PC4371X-100G
APOSPC4371X-25GEA
1540
CZK
APOSPC4371X-25G
APOSPC4371X-5G
APOSPC4371X-100G
4-Fluoro-3-(trifluoromethyl)benzamide 98%
4-Fluoro-3-(trifluoromethyl)benzamide
Vzorec:
C₈H₅F₄NO Bod tání: 84…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061269 CAS číslo: 67515-57-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...