Tisk…

ω-Chloro-3,4-difluoroacetophenone

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Chloro-3',4'-difluoroacetophenone, 2-Chloro-1-(3,4-difluorophenyl)ethanone

PC4366-1G PC4366-25G PC4366-5G
APOSPC4366-1GEA 417 CZK
APOSPC4366-1G APOSPC4366-25G APOSPC4366-5G
ω-Chloro-3,4-difluoroacetophenone
ω-Chloro-3,4-difluoroacetophenone
Vzorec: C₈H₅ClF₂O
MW: 190.5745 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03966888
CAS číslo: 51336-95-9
UN: 3261
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR