3-(Trifluoromethylthio)phenylacetic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC4365-1G
PC4365-5G
APOSPC4365-1GEA
2280
CZK
APOSPC4365-1G
APOSPC4365-5G
3-(Trifluoromethylthio)phenylacetic acid 98%
3-(Trifluoromethylthio)phenylacetic acid
Vzorec:
C₉H₇F₃O₂S MW: 236,21 g/mol Bod tání: 53…55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236350 CAS číslo: 239080-04-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...