Tisk…

N-(tert-Butoxycarbonyl)-3,4-difluoro-L-phenylalanine 95%

Dodavatel: Apollo Scientific

Synonyms: Boc-Phe(3,4-F)-OH, Boc-Phe(3,4-DiF)-OH, N-Boc-3,4-difluoro-L-phenylalanine

PC4360-10G PC4360-1G PC4360-5G
APOSPC4360-10GEA 22160 CZK
APOSPC4360-10G APOSPC4360-1G APOSPC4360-5G
N-(tert-Butoxycarbonyl)-3,4-difluoro-L-phenylalanine 95%
N-(tert-Butoxycarbonyl)-3,4-difluoro-L-phenylalanine
Vzorec: C₁₄H₁₇F₂NO₄
MW: 301,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00672518
CAS číslo: 198474-90-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR