Tisk…

Trifluorolactic acid 96%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3,3,3-Trifluoro-2-hydroxypropanoic acid

PC4338-1G PC4338-5G
APOSPC4338-1GEA 3150 CZK
APOSPC4338-1G APOSPC4338-5G
Trifluorolactic acid 96%
Trifluorolactic acid
Vzorec: C₃H₃F₃O₃
MW: 144,05 g/mol
Bod varu: 157 °C/123mmHg
Bod tání: 66…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00051802
CAS číslo: 684-07-1
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR