tert-Butyl 4-fluoro-4-(hydroxymethyl)piperidine-1-carboxylate 97%
Dodavatel: Apollo Scientific
Synonyms:
1-Boc-4-Fluoro-4-(hydroxymethyl)piperidine
PC430313-250MG
PC430313-1G
APOSPC430313-250MGEA
3150
CZK
APOSPC430313-250MG
APOSPC430313-1G
tert-Butyl 4-fluoro-4-(hydroxymethyl)piperidine-1-carboxylate 97%
tert-Butyl 4-fluoro-4-(hydroxymethyl)piperidine-1-carboxylate
Vzorec:
C₁₁H₂₀FNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD08062513 CAS číslo: 614730-97-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...