N-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-(2-Nitro-4-(trifluoromethyl)phenyl)piperidine
PC4302-1G
PC4302-5G
APOSPC4302-1GEA
622
CZK
APOSPC4302-1G
APOSPC4302-5G
N-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine 97%
N-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine
Vzorec:
C₁₂H₁₃F₃N₂O₂ MW: 274,240 g/mol Bod tání: 48…50 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00052908 CAS číslo: 1692-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...