Tisk…

2-Cyano-4-(trifluoromethyl)phenylboronic acid pinacol ester

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)benzonitrile

PC4283-5G
APOSPC4283-5GEA 5530 CZK
APOSPC4283-5G
2-Cyano-4-(trifluoromethyl)phenylboronic acid pinacol ester
2-Cyano-4-(trifluoromethyl)phenylboronic acid pinacol ester
Vzorec: C₁₄H₁₅BF₃NO₂
MW: 297,0806 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878539
CAS číslo: 1073355-21-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR