4-Amino-1,2-di(trifluoromethyl)benzene 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC4223-1G
PC4223-250MG
APOSPC4223-1GEA
12400
CZK
APOSPC4223-1G
APOSPC4223-250MG
4-Amino-1,2-di(trifluoromethyl)benzene 98%
4-Amino-1,2-di(trifluoromethyl)benzene
Vzorec:
C₈H₅F₆N Bod varu: 104…106 °C/11mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD01312619 CAS číslo: 2965-07-3 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...