Tisk…

2-Bromo-3-(trifluoromethoxy)pyridine 95%

Dodavatel: Apollo Scientific
Varování

PC421055-1G PC421055-250MG PC421055-5G
APOSPC421055-1GEA 14090 CZK
APOSPC421055-1G APOSPC421055-250MG APOSPC421055-5G
2-Bromo-3-(trifluoromethoxy)pyridine 95%
2-Bromo-3-(trifluoromethoxy)pyridine
Vzorec: C₆H₃BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD20486158
CAS číslo: 1206978-11-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR