2-Chloro-4-(difluoromethoxy)pyridine
Dodavatel: Apollo Scientific
|
Varování
|
PC421026-1G
PC421026-250MG
PC421026-5G
APOSPC421026-1GEA
2980
CZK
APOSPC421026-1G
APOSPC421026-250MG
APOSPC421026-5G
2-Chloro-4-(difluoromethoxy)pyridine
2-Chloro-4-(difluoromethoxy)pyridine
Vzorec:
C₆H₄ClF₂NO Storage Temperature: Ambient |
MDL Number:
MFCD13185848 CAS číslo: 1206978-15-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...