1-(Perfluoro-n-hexyl)tetradecane
Dodavatel: Apollo Scientific
|
Varování
|
PC4208-1G
PC4208-5G
APOSPC4208-1GEA
3940
CZK
APOSPC4208-1G
APOSPC4208-5G
1-(Perfluoro-n-hexyl)tetradecane
1-(Perfluoro-n-hexyl)tetradecane
Vzorec:
C₂₀H₂₉F₁₃ Storage Temperature: Ambient |
MDL Number:
MFCD04038341 CAS číslo: 154628-00-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...