(2-Chloro-4,5-difluorophenyl)acetic acid 98%
Dodavatel: Apollo Scientific
PC4193-1G
PC4193-5G
PC4193-10G
APOSPC4193-1GEA
3310
CZK
APOSPC4193-1G
APOSPC4193-5G
APOSPC4193-10G
(2-Chloro-4,5-difluorophenyl)acetic acid 98%
(2-Chloro-4,5-difluorophenyl)acetic acid
Vzorec:
C₈H₅ClF₂O₂ MW: 206,58 g/mol Bod tání: 99…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD04973421 CAS číslo: 883502-00-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...