Tisk…

3'-Fluoropropiophenone 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: m-Fluoropropiophenone, 1-(3-Fluorophenyl)propan-1-one

PC4189-100G PC4189-25G PC4189-5G
APOSPC4189-100GEA 2400 CZK
APOSPC4189-100G APOSPC4189-25G APOSPC4189-5G
3'-Fluoropropiophenone 97%
3'-Fluoropropiophenone
Vzorec: C₉H₉FO
Bod varu: 94…96 °C/5mmHg
Storage Temperature: Ambient
MDL Number: MFCD00061245
CAS číslo: 455-67-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR