Tisk…

4-(2-Hydroxyhexafluoroisopropyl)benzoic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(1,1,1,3,3,3-Hexafluoro-2-hydroxyprop-2-yl)benzoic acid

PC4182-1G PC4182-5G PC4182-25G
APOSPC4182-1GEA 1870 CZK
APOSPC4182-1G APOSPC4182-5G APOSPC4182-25G
4-(2-Hydroxyhexafluoroisopropyl)benzoic acid 97%
4-(2-Hydroxyhexafluoroisopropyl)benzoic acid
Vzorec: C₁₀H₆F₆O₃
MW: 288,15 g/mol
Bod tání: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00236043
CAS číslo: 16261-80-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR