3-Fluorophthalic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC4179L-5G
PC4179L-1G
PC4179L-25G
APOSPC4179L-5GEA
827
CZK
APOSPC4179L-5G
APOSPC4179L-1G
APOSPC4179L-25G
3-Fluorophthalic acid 98%
3-Fluorophthalic acid
Vzorec:
C₈H₅FO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00039524 CAS číslo: 1583-67-1 EINECS: 216-433-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...