Tisk…

3-Fluorophenyl isocyanate 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Fluorophenylisocyanate, 1-Fluoro-3-isocyanatobenzene

PC4165-25G PC4165-5G
APOSPC4165-25GEA 2150 CZK
APOSPC4165-25G APOSPC4165-5G
3-Fluorophenyl isocyanate 97%
3-Fluorophenyl isocyanate

  • May form small amount of sediment.

Vzorec: C₇H₄FNO
MW: 137,11 g/mol
Bod varu: 56…58 °C/14mmHg
Hustota: 1,201 g/cm³ (25 °C)
Bod vzplanutí: 41 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002015
CAS číslo: 404-71-7
EINECS: 206-966-1
UN: 3080
ADR: 6.1+3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR