2-Fluorophenylhydrazinium chloride 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(2-Fluorophenyl)hydrazine hydrochloride, (2-Fluorphenyl)hydrazin HCl, 1-(2-Fluorophenyl)hydrazine hydrochloride, 2-Fluorophenylhydrazine hydrochloride
PC4152-100G
PC4152-25G
APOSPC4152-100GEA
2150
CZK
APOSPC4152-100G
APOSPC4152-25G
2-Fluorophenylhydrazinium chloride 98%
2-Fluorophenylhydrazinium chloride
Vzorec:
C₆H₈ClFN₂ MW: 162,59 g/mol Bod tání: 200…205 °C (rozkl.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00012927 CAS číslo: 2924-15-4 EINECS: 220-885-9 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...