Tisk…

1-(2-Fluorophenyl)biguanide hydrochloride 97%

Dodavatel: Apollo Scientific
Varování

PC4149G-1G PC4149G-5G PC4149G-25G
APOSPC4149G-1GEA 1450 CZK
APOSPC4149G-1G APOSPC4149G-5G APOSPC4149G-25G
1-(2-Fluorophenyl)biguanide hydrochloride 97%
1-(2-Fluorophenyl)biguanide hydrochloride
Vzorec: C₈H₁₁ClFN₅
Bod tání: 237…241 °C
Storage Temperature: Ambient
MDL Number: MFCD00236682
CAS číslo: 66088-51-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR