Tisk…

2-Fluoro-4-biphenylboronic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Fluoro-4-biphenylylboronic acid, (3-fluoro-4-phenyl)phenylboronic acid, 3-Fluoro-4-phenylbenzeneboronic acid, 2-Fluoro-1,1'-biphenyl-4-ylboronic acid

PC4135-1G PC4135-25G PC4135-5G
APOSPC4135-1GEA 622 CZK
APOSPC4135-1G APOSPC4135-25G APOSPC4135-5G
2-Fluoro-4-biphenylboronic acid 97%
2-Fluoro-4-biphenylboronic acid
Vzorec: C₁₂H₁₀BFO₂
MW: 216,02 g/mol
Bod tání: 242…250 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01075707
CAS číslo: 178305-99-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR