Tisk…

1-[Chloro(difluoro)methoxy]-4-nitrobenzene 97%

Dodavatel: Apollo Scientific
Varování

PC4134-1G PC4134-500MG PC4134-5G
APOSPC4134-1GEA 1660 CZK
APOSPC4134-1G APOSPC4134-500MG APOSPC4134-5G
1-[Chloro(difluoro)methoxy]-4-nitrobenzene 97%
1-[Chloro(difluoro)methoxy]-4-nitrobenzene
Vzorec: C₇H₄ClF₂NO₃
MW: 223,56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01820808
CAS číslo: 40750-71-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR