8-(Trifluoromethyl)quinoline-6-boronic acid
Dodavatel: Apollo Scientific
Synonyms:
(8-(Trifluoromethyl)quinolin-6-yl)boronic acid
PC412567-1G
PC412567-250MG
APOSPC412567-1GEA
11580
CZK
APOSPC412567-1G
APOSPC412567-250MG
8-(Trifluoromethyl)quinoline-6-boronic acid
8-(Trifluoromethyl)quinoline-6-boronic acid
Vzorec:
C₁₀H₇BF₃NO₂ MW: 240,9743 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11504852 CAS číslo: 1072951-47-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...