Tisk…

5-(Trifluoromethyl)-2-pyridineboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: (5-(Trifluoromethyl)pyridin-2-yl)boronic acid

PC412562-1G PC412562-250MG
APOSPC412562-1GEA 10490 CZK
APOSPC412562-1G APOSPC412562-250MG
5-(Trifluoromethyl)-2-pyridineboronic acid
5-(Trifluoromethyl)-2-pyridineboronic acid
Vzorec: C₆H₅BF₃NO₂
MW: 190,9156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06227445
CAS číslo: 1162257-58-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR