Tisk…

2-(Trifluoromethyl)phenylboronic acid pinacol ester 98%

Dodavatel: Apollo Scientific

Synonyms: 4,4,5,5-Tetramethyl-2-(2-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane

PC412560-1G PC412560-25G PC412560-5G
APOSPC412560-1GEA 2900 CZK
APOSPC412560-1G APOSPC412560-25G APOSPC412560-5G
2-(Trifluoromethyl)phenylboronic acid pinacol ester 98%
2-(Trifluoromethyl)phenylboronic acid pinacol ester
Vzorec: C₁₃H₁₆BF₃O₂
MW: 272.0712 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06795676
CAS číslo: 1073339-21-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR