Tisk…
2-Hydroxy-4-(trifluoromethyl)phenylboronic acid 97%
Dodavatel: Apollo Scientific
Synonyms: (2-Hydroxy-4-(trifluoromethyl)phenyl)boronic acid
PC412500-1G PC412500-500MG PC412500-5G PC412500-25G
APOSPC412500-1GEA 2480 CZK
APOSPC412500-1G APOSPC412500-500MG APOSPC412500-5G APOSPC412500-25G
2-Hydroxy-4-(trifluoromethyl)phenylboronic acid 97%
2-Hydroxy-4-(trifluoromethyl)phenylboronic acid
Vzorec: C₇H₆BF₃O₃
MW: 205,9270 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094996
CAS číslo: 1072951-50-8