2-Fluoro-5-pyridineboronic acid pinacol ester
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(6-Fluoro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Fluoropyridine-3-boronic acid Pinacol ester, 2-Fluoropyridine-5-boronic acid pinacol ester
PC412477-10G
PC412477-1G
PC412477-5G
APOSPC412477-10GEA
1580
CZK
APOSPC412477-10G
APOSPC412477-1G
APOSPC412477-5G
2-Fluoro-5-pyridineboronic acid pinacol ester
2-Fluoro-5-pyridineboronic acid pinacol ester
Vzorec:
C₁₁H₁₅BFNO₂ MW: 223.05 Bod tání: 29…33 °C Storage Temperature: Ambient |
MDL Number:
MFCD03412792 CAS číslo: 444120-95-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...