Tisk…

2-Fluoro-3-isobutoxyphenylboronic acid

Dodavatel: Apollo Scientific

PC412432-1G PC412432-250MG
APOSPC412432-1GEA 7250 CZK
APOSPC412432-1G APOSPC412432-250MG
2-Fluoro-3-isobutoxyphenylboronic acid
2-Fluoro-3-isobutoxyphenylboronic acid
Vzorec: C₁₀H₁₄BFO₃
Storage Temperature: Ambient
MDL Number: MFCD12546465
CAS číslo: 1217500-66-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR