Tisk…

4-(2'-Fluorobenzyloxy)phenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

PC412405-25G PC412405-5G
APOSPC412405-25GEA 21490 CZK
APOSPC412405-25G APOSPC412405-5G
4-(2'-Fluorobenzyloxy)phenylboronic acid 98%
4-(2'-Fluorobenzyloxy)phenylboronic acid
Vzorec: C₁₃H₁₂BFO₃
Storage Temperature: Ambient
MDL Number: MFCD08705274
CAS číslo: 1072951-78-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR