Tisk…

2,6-Difluoro-4-pyridineboronic acid pinacol ester

Dodavatel: Apollo Scientific

PC412384-1G PC412384-500MG
APOSPC412384-1GEA 3540 CZK
APOSPC412384-1G APOSPC412384-500MG
2,6-Difluoro-4-pyridineboronic acid pinacol ester
2,6-Difluoro-4-pyridineboronic acid pinacol ester
Vzorec: C₁₁H₁₄BF₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD12913987
CAS číslo: 1204333-58-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR