Tisk…

2-Cyano-4-fluorophenylboronic acid 95+%

Dodavatel: Apollo Scientific

Synonyms: (2-Cyano-4-fluorophenyl)boronic acid

PC412334-1G PC412334-250MG
APOSPC412334-1GEA 2360 CZK
APOSPC412334-1G APOSPC412334-250MG
2-Cyano-4-fluorophenylboronic acid 95+%
2-Cyano-4-fluorophenylboronic acid
Vzorec: C₇H₅BFNO₂
MW: 164,9295 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546526
CAS číslo: 876601-43-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR