Tisk…

2-Chloro-4-(trifluoromethoxy)phenylboronic acid 98%

Dodavatel: Apollo Scientific

Synonyms: (2-Chloro-4-(trifluoromethoxy)phenyl)boronic acid, 2-Chloro-4-trifluoromethoxyphenylboronic acid

PC412323-1G PC412323-250MG PC412323-5G
APOSPC412323-1GEA 5530 CZK
APOSPC412323-1G APOSPC412323-250MG APOSPC412323-5G
2-Chloro-4-(trifluoromethoxy)phenylboronic acid 98%
2-Chloro-4-(trifluoromethoxy)phenylboronic acid
Vzorec: C₇H₅BClF₃O₃
MW: 240,3720 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11040272
CAS číslo: 345226-20-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR